Learn More
AnaSpec OPA
Catalog No. 50850600
Encompass
OPA[[o-Phthaldialdehyde] *UltraPure Grade*] ; MW 134.1; (1 g) Fluorogenic indicator for primary amino group; Widely used for quantitation of peptides and proteins.
Catalog No. 50-850-600 Supplier AnaSpec Supplier No. AS83012
OPA[[o-Phthaldialdehyde] *UltraPure Grade*] ; MW 134.1; (1 g) Fluorogenic indicator for primary amino group; Widely used for quantitation of peptides and proteins.