Learn More
Pickering Laboratories Inc o-Phthalaldehyde, 5 g
SDP
Catalog No. NC9147343
Encompass_Preferred
Chromatographic Grade™ For Automated Post-column Derivatization of Primary Amines. Primary amines form highly fluorescent adducts when reacted with o-Phthalaldehyde (OPA) and a mercaptan under basic conditions. The products of this reaction, 1-alkyl-2-thioalkyl-substituted isoindoles, exhibit optimal excitation at 330 nm and maximal emission at 465 nm.
Catalog No. NC9147343 Supplier Pickering Laboratories Inc Supplier No. O120
Chromatographic Grade™ For Automated Post-column Derivatization of Primary Amines. Primary amines form highly fluorescent adducts when reacted with o-Phthalaldehyde (OPA) and a mercaptan under basic conditions. The products of this reaction, 1-alkyl-2-thioalkyl-substituted isoindoles, exhibit optimal excitation at 330 nm and maximal emission at 465 nm.