Learn More
Pickering Laboratories Inc OPA DILUENT 4X950ML CASE
SDP
Supplier:  Pickering Laboratories Inc CB910
Encompass_Preferred
o-Phthalaldehyde (OPA) Diluent for Carbamate Analysis, case of 4 (950 mL /bottle) EPA 531.1 and 531.2
Catalog No. NC9116804