Learn More
Aqua Solutions OPA Solution
SDP
Catalog No. NC1802468
Encompass_Preferred
OPA Solution | 1.17% Methanol (67-56-1) 0.749% Boric Acid ( 10043-35-3) 0.449% Sodium Hydroxide (1310-73-2) 0.0197% Phthalic Dicarboxaldehyde (643-79-8) 97.612% Water (7732-18-5)
This item is not returnable. View return policy
Catalog No. NC1802468 Supplier Aqua Solutions Supplier No. MLU014
OPA Solution | 1.17% Methanol (67-56-1) 0.749% Boric Acid ( 10043-35-3) 0.449% Sodium Hydroxide (1310-73-2) 0.0197% Phthalic Dicarboxaldehyde (643-79-8) 97.612% Water (7732-18-5)