Learn More
Aviva Systems Biology Corp OPA1 Antibody - N-terminal region (ARP57715_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57715_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OPA1. It was validated on Western Blot. . Alternate names: FLJ12460, KIAA0567, MGM1, NPG, NTG, largeGApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-359