Learn More
TARGETMOL CHEMICALS INC OPAGANIB 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T675010MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Opaganib (ABC294640) is an orally active and specific sphingosine kinase-2 (SphK2) inhibitor (IC50 60 uM). purity: 99%
Catalog No. 50-257-6462