Learn More
Quanterix Inc OPAL 520 REAGENT PACK
SDP
Catalog No. NC1601877
Encompass
Opal™ is a practical workflow for simultaneous detection of up to 6 tissue biomarkers plus nuclear counterstain, within a single image. The method is similar to standard immunohistochemistry and is accessible to many laboratories where standard IHC method development is performed. Standalone Opal Fluorophore reagent packs allow you to pick and choose your panel's biomarker quantity and design.
This item is not returnable. View return policy
Catalog No. NC1601877 Supplier Quanterix Inc Supplier No. FP1487001KT
Opal™ is a practical workflow for simultaneous detection of up to 6 tissue biomarkers plus nuclear counterstain, within a single image. The method is similar to standard immunohistochemistry and is accessible to many laboratories where standard IHC method development is performed. Standalone Opal Fluorophore reagent packs allow you to pick and choose your panel's biomarker quantity and design.