Learn More
Quanterix Inc OPAL Fluorophore Reagent Packs
SDP
Catalog No. NC1601878
Encompass
Akoya Biosciences' Opal Fluorophore Reagent Packs contain 7 distinct fluorophores for multiplex immunofluorescence, enabling simultaneous staining with up to 7 markers. These photostable dyes work with formalin-fixed samples and standard fluorescence microscopes, facilitating high-plex tissue analysis.
This item is not returnable. View return policy
Catalog No. NC1601878 Supplier Quanterix Inc Supplier No. FP1488001KT
Akoya Biosciences' Opal Fluorophore Reagent Packs contain 7 distinct fluorophores for multiplex immunofluorescence, enabling simultaneous staining with up to 7 markers. These photostable dyes work with formalin-fixed samples and standard fluorescence microscopes, facilitating high-plex tissue analysis.