Learn More
Quanterix Inc Opal Polaris 780 Reagent Pack
SDP
Catalog No. NC1685841
Encompass
Opal™ is a practical workflow for simultaneous detection of up to 8 tissue biomarkers plus nuclear counterstain, within a single image. The method is similar to standard immunohistochemistry and is accessible to many laboratories where standard IHC method development is performed.
Catalog No. NC1685841 Supplier Quanterix Inc Supplier No. FP1501001KT
Opal™ is a practical workflow for simultaneous detection of up to 8 tissue biomarkers plus nuclear counterstain, within a single image. The method is similar to standard immunohistochemistry and is accessible to many laboratories where standard IHC method development is performed.