Learn More
Glass Expansion OpalMist DC Nebulizer 0.1mL/min, Includes NFT-025 UniFit Connector with 0.25mm ID x 1.3mm OD x 700mm long tubing (1 piece) and 70-803-0969 Gas Fitting with Ratchet Nebulizer connector
SDP
Catalog No. 501907372
Encompass_Preferred
OpalMist DC Nebulizer 0.1mL/min, includes UniFit Connector with 0.25mm ID x 1.3mm OD x 700mm long tubing (1 pc) & DC Gas Fitting Type 11 with ratchet connector (1 pc). Compatible with Analytik Jena ICP-MS and Leeman.Optimum nebulizer gas flow rate is 1.0L/min & sample flow rate is 0.1mL/min. Recommended not to exceed optimum 0.1mL/min sample flow rate; however, lower gas & sample flow rates are OK. The backpressure should read ~40 psi at 1.0L/min gas flow rate. OpalMist is made from high purity PFA resulting in the lowest background & exceptional resistance to HF. Tolerates up to 15% TDS & 75um particulates.Replacement parts include P/Ns NFT-025 & 502703076.Recommended nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) & Eluo Nebulizer Cleaning Tool (P/N 501907353). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7372 Supplier Glass Expansion Supplier No. A111PFA01
OpalMist DC Nebulizer 0.1mL/min, includes UniFit Connector with 0.25mm ID x 1.3mm OD x 700mm long tubing (1 pc) & DC Gas Fitting Type 11 with ratchet connector (1 pc). Compatible with Analytik Jena ICP-MS and Leeman.Optimum nebulizer gas flow rate is 1.0L/min & sample flow rate is 0.1mL/min. Recommended not to exceed optimum 0.1mL/min sample flow rate; however, lower gas & sample flow rates are OK. The backpressure should read ~40 psi at 1.0L/min gas flow rate. OpalMist is made from high purity PFA resulting in the lowest background & exceptional resistance to HF. Tolerates up to 15% TDS & 75um particulates.Replacement parts include P/Ns NFT-025 & 502703076.Recommended nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) & Eluo Nebulizer Cleaning Tool (P/N 501907353). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001