Learn More
Glass Expansion OpalMist Nebulizer 0.4mL/min & 0.5 x 1.6 x 700mm Tube, Includes NFT-16-50 UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 piece), EL-1 EzyLok Connector Kit, and 70-803-0911 Ratchet EzyLok Connector.
SDP
Catalog No. 501907540
Encompass_Preferred
OpalMist U-Series Nebulizer 0.4mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 pc) & EzyLok Connector (1 pc).Optimum nebulizer gas flow rate is 1.0L/min & sample flow rate is 0.4mL/min. Recommended not to exceed optimum 0.4mL/min sample flow rate; however, lower gas & sample flow rates are OK. The backpressure should read ~40 psi at 1.0L/min gas flow rate. OpalMist is made from high purity PFA resulting in the lowest background & exceptional resistance to HF. Tolerates up to 15% TDS & 75um particulates.Replacement parts include P/Ns NFT-16-50, 502702994 & 501907289.Recommended nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) & Eluo Nebulizer Cleaning Tool (P/N 501907353). When nebulizer is not in use, store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) OpalMist to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7540 Supplier Glass Expansion Supplier No. ARG1PFA04
OpalMist U-Series Nebulizer 0.4mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 pc) & EzyLok Connector (1 pc).Optimum nebulizer gas flow rate is 1.0L/min & sample flow rate is 0.4mL/min. Recommended not to exceed optimum 0.4mL/min sample flow rate; however, lower gas & sample flow rates are OK. The backpressure should read ~40 psi at 1.0L/min gas flow rate. OpalMist is made from high purity PFA resulting in the lowest background & exceptional resistance to HF. Tolerates up to 15% TDS & 75um particulates.Replacement parts include P/Ns NFT-16-50, 502702994 & 501907289.Recommended nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) & Eluo Nebulizer Cleaning Tool (P/N 501907353). When nebulizer is not in use, store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) OpalMist to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001