Learn More
Glass Expansion OpalMist Nebulizer 2mL/min, Includes NFT-16-75 UniFit Connector with 0.75mm ID x 1.6mm OD x 700mm long tubing (1 piece), EL-1 EzyLok Connector Kit, and 70-803-0911 Ratchet EzyLok Connector
SDP
Catalog No. 501907520
Encompass_Preferred
OpalMist U-Series Nebulizer 2.0mL/min, includes UniFit Connector with 0.75mm ID x 1.6mm OD x 700mm long tubing (1 pc) & EzyLok Connector (1 pc).Optimum nebulizer gas flow rate is 0.7L/min & sample flow rate is 2.0mL/min. Recommended not to exceed optimum 2.0mL/min sample flow rate; however, lower gas & sample flow rates are OK. The backpressure should read ~40 psi at 0.7L/min gas flow rate. OpalMist is made from high purity PFA resulting in the lowest background & exceptional resistance to HF. Tolerates up to 15% TDS & 75um particulates.Replacement parts include P/Ns NFT-16-75, 502702994 & 501907289.Recommended nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) & Eluo Nebulizer Cleaning Tool (P/N 501907353). When nebulizer is not in use, store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) OpalMist to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7520 Supplier Glass Expansion Supplier No. ARG07PFA2
OpalMist U-Series Nebulizer 2.0mL/min, includes UniFit Connector with 0.75mm ID x 1.6mm OD x 700mm long tubing (1 pc) & EzyLok Connector (1 pc).Optimum nebulizer gas flow rate is 0.7L/min & sample flow rate is 2.0mL/min. Recommended not to exceed optimum 2.0mL/min sample flow rate; however, lower gas & sample flow rates are OK. The backpressure should read ~40 psi at 0.7L/min gas flow rate. OpalMist is made from high purity PFA resulting in the lowest background & exceptional resistance to HF. Tolerates up to 15% TDS & 75um particulates.Replacement parts include P/Ns NFT-16-75, 502702994 & 501907289.Recommended nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) & Eluo Nebulizer Cleaning Tool (P/N 501907353). When nebulizer is not in use, store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) OpalMist to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001