Learn More
Alkali Scientific Western Blot Box, Removable Lift Off Lid, for Novex Minigel 8.6 X 8.6 X 2.8cm, Opaque Black, 10/pk
SDP
Catalog No. 502389142
Encompass_Preferred
Western blot boxes and containers are opaque black for light sensitive samples. They fit nearly any sized blot and feature a hinged lid that easy to open and close. The corners are 90˚ and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot boxes. When you match the size of the box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.
Catalog No. 50-238-9142 Supplier Alkali Scientific Supplier No. BC13008BK
Western blot boxes and containers are opaque black for light sensitive samples. They fit nearly any sized blot and feature a hinged lid that easy to open and close. The corners are 90˚ and they have a flatter and more uniform surface. Use less reagents and antibodies with these cost-efficient western blot boxes. When you match the size of the box with your blot you're able to fit more blots on your rocker and shaker allowing for a smaller blot footprint.