Learn More
Research Products International Corp Opaque Thin Wall PCR Plates, RNase and DNase Free, Black, 50 per Case
SDP
Catalog No. 50212250
Encompass
Opaque black color ensures the lowest level of background fluorescence. Thin wall plate design provides rapid and uniform thermal transfer. Skirted base allows easier handling and provides area for bar coding and labeling. Features alpha/numeric grid index and notched corner for quick orientation. Can be sealed with adhesive films. Made of heavy duty polypropylene and are autoclavable.
The plates are packaged in tamperproof resealable bags of 10 plates, 50 plates per case.
Catalog No. 50-212-250 Supplier Research Products International Corp Supplier No. 141351
Opaque black color ensures the lowest level of background fluorescence. Thin wall plate design provides rapid and uniform thermal transfer. Skirted base allows easier handling and provides area for bar coding and labeling. Features alpha/numeric grid index and notched corner for quick orientation. Can be sealed with adhesive films. Made of heavy duty polypropylene and are autoclavable.
The plates are packaged in tamperproof resealable bags of 10 plates, 50 plates per case.