Learn More
TARGETMOL CHEMICALS INC OPAVIRALINE 5MG
SDP
Catalog No. 502580532
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Opaviraline (GW-420867X) is a potent reverse transcriptase inhibitor that inhibits Human immunodeficiency virus 1 and has the potential to treat HIV infection. purity: 100%
Catalog No. 50-258-0532 Supplier TARGETMOL CHEMICALS INC Supplier No. T282555MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Opaviraline (GW-420867X) is a potent reverse transcriptase inhibitor that inhibits Human immunodeficiency virus 1 and has the potential to treat HIV infection. purity: 100%