Learn More
Selleck Chemical LLC OPB-171775 E4695-25MG
SDP
Catalog No. 5000038993
Encompass_Preferred
OPB-171775 functions as a molecular glue promoting the formation of a complex between phosphodiesterase 3A (PDE3A) and Schlafen family member 12 (SLFN12) This interaction triggers multiple cellular stresses ultimately leading to cell death It also demonstrates significant anticancer activity in patient-derived xenograft (PDX) models of Gastrointestinal stromal tumor (GIST)
Catalog No. 50-000-38993 Supplier Selleck Chemical LLC Supplier No. E469525MG
OPB-171775 functions as a molecular glue promoting the formation of a complex between phosphodiesterase 3A (PDE3A) and Schlafen family member 12 (SLFN12) This interaction triggers multiple cellular stresses ultimately leading to cell death It also demonstrates significant anticancer activity in patient-derived xenograft (PDX) models of Gastrointestinal stromal tumor (GIST)