Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OPC-14523 hydrochlor 10mM 1mL | 145969-31-9 | 450.40 g/mol | C23H29Cl2N3O2 | 1 ML
SDP

Catalog No. 5000308232
Encompass_Preferred

OPC-14523 hydrochloride is a sigma and 5-HT1A receptor agonist supplied as a hydrochloride salt for research use. It is available as a lyophilized solid and as a ready-to-use 10 mM solution in DMSO, with documented solubility and storage recommendations for in vitro and in vivo studies.

  • Hydrochloride salt form for improved stability and handling.
  • High purity (99.95%) suitable for pharmacology studies.
  • Chemical formula C23H29Cl2N3O2; molecular weight 450.40 g/mol.
  • Available as solid in multiple mg sizes and as a 10 mM, 1 mL solution in DMSO.
  • Soluble in DMSO (~83.3 mg/mL) and moderately soluble in water with warming.
  • Recommended storage: sealed, away from moisture; in solvent: -20°C short term, -80°C long term.

Catalog No. 50-003-08232 Supplier Medchemexpress LLC Supplier No. HY116594A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OPC-14523 hydrochloride is a sigma and 5-HT1A receptor agonist supplied as a hydrochloride salt for research use. It is available as a lyophilized solid and as a ready-to-use 10 mM solution in DMSO, with documented solubility and storage recommendations for in vitro and in vivo studies.

  • Hydrochloride salt form for improved stability and handling.
  • High purity (99.95%) suitable for pharmacology studies.
  • Chemical formula C23H29Cl2N3O2; molecular weight 450.40 g/mol.
  • Available as solid in multiple mg sizes and as a 10 mM, 1 mL solution in DMSO.
  • Soluble in DMSO (~83.3 mg/mL) and moderately soluble in water with warming.
  • Recommended storage: sealed, away from moisture; in solvent: -20°C short term, -80°C long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.