Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OPC 4392 hydrochloride | 1329509-60-5 | 99.0% | 427.97 | C24H30ClN3O2 | 5 MG
SDP

Catalog No. 5000242037
Encompass_Preferred

OPC 4392 hydrochloride is the hydrochloride salt of a dopamine receptor modulator described as a presynaptic dopamine receptor agonist and a postsynaptic D2 antagonist. It is intended for laboratory research and preclinical studies and is supplied in small milligram pack sizes.

  • High purity (98.99%) for reliable experimental results.
  • Hydrochloride salt form for improved solubility in DMSO.
  • Described pharmacology: presynaptic dopamine agonist and postsynaptic D2 antagonist.
  • Molecular weight 427.97 and formula C24H30ClN3O2 for accurate dosing calculations.
  • Supplied in small milligram pack sizes suitable for in vitro testing.
  • CAS registry number 1329509-60-5 for unambiguous identification.

Catalog No. 50-002-42037 Supplier Medchemexpress LLC Supplier No. HY106781A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OPC 4392 hydrochloride is the hydrochloride salt of a dopamine receptor modulator described as a presynaptic dopamine receptor agonist and a postsynaptic D2 antagonist. It is intended for laboratory research and preclinical studies and is supplied in small milligram pack sizes.

  • High purity (98.99%) for reliable experimental results.
  • Hydrochloride salt form for improved solubility in DMSO.
  • Described pharmacology: presynaptic dopamine agonist and postsynaptic D2 antagonist.
  • Molecular weight 427.97 and formula C24H30ClN3O2 for accurate dosing calculations.
  • Supplied in small milligram pack sizes suitable for in vitro testing.
  • CAS registry number 1329509-60-5 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.