Learn More
Sigma Aldrich Fine Chemicals Biosciences o-Phenylenediamine dihydrochloride, 615-28-1, MFCD00012966, 30 mg
Catalog No. NC0080253
Encompass
Linear Formula: C6H4(NH2)2·2HCl, Molecular Weight: 181.06, tablet, 30 mg substrate per tablet. o-Phenylenediamine dihydrochloride has been used as a substrate for horse radish peroxidase in detecting: human immunoglobulin, CTLA4Ig in serum samples, and albumin in culture medium.
Catalog No. NC0080253 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P841250TAB
Linear Formula: C6H4(NH2)2·2HCl, Molecular Weight: 181.06, tablet, 30 mg substrate per tablet. o-Phenylenediamine dihydrochloride has been used as a substrate for horse radish peroxidase in detecting: human immunoglobulin, CTLA4Ig in serum samples, and albumin in culture medium.