Learn More
Waters Corp Open Access Valve Cartridge, 6 Port, 2 Position
Catalog No. 502391776 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement Open Access Valve Cartridge features a 6 port, 2 position design. It is specifically used with a 2777 Sample Manager in conjunction with ACQUITY Solvent Managers and an Everflow Valve.
  • Replacement valve cartridge with 6 ports and 2 positions.
  • Designed for use with a 2777 Sample Manager.
  • Compatible with ACQUITY Solvent Managers.
  • Compatible with an Everflow Valve.
Catalog No. 50-239-1776 Supplier Waters Corp Supplier No. 700011083
Add to Cart
Add to Cart
Request a Quote
This replacement Open Access Valve Cartridge features a 6 port, 2 position design. It is specifically used with a 2777 Sample Manager in conjunction with ACQUITY Solvent Managers and an Everflow Valve.
  • Replacement valve cartridge with 6 ports and 2 positions.
  • Designed for use with a 2777 Sample Manager.
  • Compatible with ACQUITY Solvent Managers.
  • Compatible with an Everflow Valve.