Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Open Case Form Electroscope
GREENER_CHOICESCI_ED

Catalog No. S139444 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139444 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139444 Supplier Supertek™ Supplier No. PH45265
Add to Cart
Add to Cart
This item is not returnable. View return policy

Charge is indicated by deflection of an aluminized plastic straw which rotates freely on the saddle of the conducting rod within this Supertek™ Open Case Form Electroscope.

  • A scale on the instruments back is graduated in degrees to allow measurement of the deflection.
  • The conducting rod is recessed in the metal housing to shield against air currents.
  • Comes with replacement aluminized straws.
  • Instructions included.
  • Grades 6+

Specifications

Grade 6+
Format Kit
Languages English
Subject Physical Science - Electricity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.