Learn More
Grainger Open End Rack 19.88 L PK3
Catalog No. 502750914
Encompass_Preferred
The Grainger Open End Rack is a 19" rack with 88 rack units of usable space. It is constructed of steel with a black powder coat finish and features fully open sides for easy access. The rack has adjustable mounting rails and includes hardware for assembly. It can support up to 1000 lbs of equipment. The overall dimensions are 24"W x 41"D x 88"H. This rack provides an open frame solution for mounting audio, video, networking and other 19" rack equipment.
Catalog No. 50-275-0914 Supplier Grainger Supplier No. 14D018
Add to Cart
Add to Cart
The Grainger Open End Rack is a 19" rack with 88 rack units of usable space. It is constructed of steel with a black powder coat finish and features fully open sides for easy access. The rack has adjustable mounting rails and includes hardware for assembly. It can support up to 1000 lbs of equipment. The overall dimensions are 24"W x 41"D x 88"H. This rack provides an open frame solution for mounting audio, video, networking and other 19" rack equipment.