Learn More
Proto™ Open End Wrenches
Catalog No. 19313496
Click to view available options
Size:
9/16 in., 5/8 in.
Catalog No. Size
19-313-496 9/16 in., 5/8 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-313-496 Supplier Proto™ Supplier No. 5773030
This item is not returnable. View return policy
Anti-Slip Design (ASD) wrench has a locking groove preventing slippage and knuckle injury.
  • Increased torque applied to fastener with fewer rounded edges
  • Unique arc design increases surface contact
Specifications
Length (English) 7 5/8 in.
Material Forged Alloy Steel
Size 9/16 in., 5/8 in.
Color Chrome
Includes Wrench
Weight (English) 0.28 lb.
Width (English) 1-17/64 in.