Learn More
MSC Proto Socket, 3/8 in Drive, 15 mm 12-Point, 1 ea
Catalog No. 501920586
Encompass
Proto 12-point deep sockets are designed for high-performance use on stubborn, damaged, or soft fasteners. Their unique TorquePlus™ configuration ensures efficient engagement, extended fastener life, and minimizes rounding.
  • Ideal for use on stubborn or damaged fasteners
  • Minimizes fastener rounding and extends fastener life
  • Allows easier turning of worn, rounded, or rusted fasteners
  • Radius-corner design for optimal stress distribution
  • Meets aerospace specification AS4984A for durability
  • Thicker nickel-chrome coating for corrosion resistance
Catalog No. 50-192-0586 Supplier MSC Supplier No. 39934161
Proto 12-point deep sockets are designed for high-performance use on stubborn, damaged, or soft fasteners. Their unique TorquePlus™ configuration ensures efficient engagement, extended fastener life, and minimizes rounding.
  • Ideal for use on stubborn or damaged fasteners
  • Minimizes fastener rounding and extends fastener life
  • Allows easier turning of worn, rounded, or rusted fasteners
  • Radius-corner design for optimal stress distribution
  • Meets aerospace specification AS4984A for durability
  • Thicker nickel-chrome coating for corrosion resistance