Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CritiCore Protective Wear Open Face Reusable Cleanroom Hood

Catalog No. 18111257
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-257 3X-Large
18-111-251 X-Small
18-111-252 Small
18-111-253 Medium
18-111-254 Large
18-111-255 X-Large
18-111-256 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 18-111-257 Supplier CritiCore Protective Wear Supplier No. 11HD3262N180013XLWHT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reusable hoods are suitable for a variety of cleanroom applications including pharmaceutical, biotechnology, and medical device manufacturing.

  • Fluid and bacteria resistant Integrity 1800™ high performance carbon striped fabric
  • Open face, split-drape design
  • Laser-cut, pre-serged seams
  • Silicon-free
  • Tunneled elastic plus horizontal and vertical adjustment snaps

Specifications

Product Type Open Face
Size 3X-Large
Material Polyester, Carbon
Color White
Closure Type Snap Front
Disposable Reusable
Latex Free Yes
Model Integrity™ 1800
Sterility Non-sterile
Includes Open Face
For Use With (Application) To Control The Migration of Particulates in Pharmaceutical Cleanroom Environments
Hood Type Open Face
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.