Learn More
Grainger Open Frame Rack 45in H 45U Steel 22 in D
Catalog No. 502752820
Encompass_Preferred
The Tripp Lite Open Frame Rack Floor Mount is a 19-inch rack mount kit that allows you to mount an open frame rack to the floor. It is made from heavy-duty steel with a durable black powder coat finish. The kit includes 2 vertical rails and 4 adjustable mounting feet to level and stabilize the rack. It supports 19-inch rack equipment up to 20U in size and 1000 lbs in weight. The open frame design allows for efficient cooling. Overall this is a sturdy, adjustable rack mounting kit for stabilizing standard 19-inch racks on the floor.
Catalog No. 50-275-2820 Supplier Grainger Supplier No. 14A334
The Tripp Lite Open Frame Rack Floor Mount is a 19-inch rack mount kit that allows you to mount an open frame rack to the floor. It is made from heavy-duty steel with a durable black powder coat finish. The kit includes 2 vertical rails and 4 adjustable mounting feet to level and stabilize the rack. It supports 19-inch rack equipment up to 20U in size and 1000 lbs in weight. The open frame design allows for efficient cooling. Overall this is a sturdy, adjustable rack mounting kit for stabilizing standard 19-inch racks on the floor.