Learn More
Grainger Open-Head Drum 30 gal Blk
Catalog No. 502749740
Encompass_Preferred
The Open Head Drum - 30 Gallon - Black is a 30 gallon open head steel drum with a rolled rim and 2 inch bung opening. It has an 18 gauge steel construction and measures 23.5 inches tall with a 22.5 inch diameter. This UN rated 1A2/Y30/S drum has a capacity of 30 gallons or 113 liters and can hold liquids up to a specific gravity of 1.9. It comes unlined and is made in the USA. This drum is ideal for storing and transporting chemicals, petroleum products, food products and other industrial liquids.
Catalog No. 50-274-9740 Supplier Grainger Supplier No. 14N864
The Open Head Drum - 30 Gallon - Black is a 30 gallon open head steel drum with a rolled rim and 2 inch bung opening. It has an 18 gauge steel construction and measures 23.5 inches tall with a 22.5 inch diameter. This UN rated 1A2/Y30/S drum has a capacity of 30 gallons or 113 liters and can hold liquids up to a specific gravity of 1.9. It comes unlined and is made in the USA. This drum is ideal for storing and transporting chemicals, petroleum products, food products and other industrial liquids.