Learn More
Grainger Approved Vendor Stock Truck, 2,000 lb, Steel, Slat Walls, 1 ea
Catalog No. 502783926
Encompass_Preferred
These metal stock trucks are engineered with slat walls enclosing three sides to provide enhanced protection against items falling from the shelves. Designed for reliability, they feature a durable welded steel construction with a protective powder-coated finish suitable for various material handling applications.
  • Solid shelf construction with fixed adjustability
  • Three slat-style side panels for item retention and visibility
  • Flush lip style for efficient loading
  • Load capacity of 2,000 lb
  • Includes four 6 in. rubber wheels in a two rigid and two swivel configuration
  • Flat handle type for easy maneuverability
  • Assembled construction for immediate use
Catalog No. 50-278-3926 Supplier Grainger Supplier No. 19G914
These metal stock trucks are engineered with slat walls enclosing three sides to provide enhanced protection against items falling from the shelves. Designed for reliability, they feature a durable welded steel construction with a protective powder-coated finish suitable for various material handling applications.
  • Solid shelf construction with fixed adjustability
  • Three slat-style side panels for item retention and visibility
  • Flush lip style for efficient loading
  • Load capacity of 2,000 lb
  • Includes four 6 in. rubber wheels in a two rigid and two swivel configuration
  • Flat handle type for easy maneuverability
  • Assembled construction for immediate use