Learn More
Ace Glass, Inc. Open ring support, PVC coated aluminum, 4in OD, 3-1/4in ID, 12in arm
SDP
Catalog No. NC0793104
Encompass
4" PVC coated open ring support has 12" extension arm for supporting glassware, such as separatory funnels and powder funnels, from the bottom. The long extension arm allows for easier connection to labframes or stands.
Catalog No. NC0793104 Supplier Ace Glass, Inc. Supplier No. 1117717
4" PVC coated open ring support has 12" extension arm for supporting glassware, such as separatory funnels and powder funnels, from the bottom. The long extension arm allows for easier connection to labframes or stands.