Learn More
Diversified Spaces™ Open-Style Wood Workbench
GSA_VASCI_ED
Catalog No. S42470 Shop All Diversified Spaces Products
Change view
Click to view available options
Dimensions (D x W x H):
54 x 64 x 31.25 in.
64 x 28 x 31.25 in.
Includes:
Four vises
No vises
Two vises
Product Type:
Workbench
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H) Includes Product Type
S42470 54 x 64 x 31.25 in. Four vises Workbench
S42462 64 x 28 x 31.25 in. No vises Workbench
S42469 54 x 64 x 31.25 in. Two vises Workbench
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S42470 Supplier Diversified Spaces™ Supplier No. WW44VM
Add to Cart
Add to Cart
Request a Quote
Features large work surface and plenty of open storage space on the maple plywood lower shelf.
  • Constructed from solid maple and maple veneer
  • 5.715cm (2.25 in.) thick maple top
  • Can be ordered with or without black vises
  • Vises need on site installation
  • SEFA compliant and MAS Certified Green
  • Finished with durable chemical resistant non-emitting UV finish
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Workbench
Color Maple
Dimensions (D x W x H) 54 x 64 x 31.25 in.
Includes Four vises
Style Open
Warranty Limited Lifetime
Material Maple