Learn More
Spectrocell Inc OPEN TOP QUARTZ 10MM CELL
Catalog No. NC1520487
Encompass
Open top quartz 10 mm path length FUV cell with PTFE lid and 2 polished windows.3.5ml. Type 1, Spectral Range: 170-2200 nm
Catalog No. NC1520487 Supplier Spectrocell Inc Supplier No. R3010
Open top quartz 10 mm path length FUV cell with PTFE lid and 2 polished windows.3.5ml. Type 1, Spectral Range: 170-2200 nm