Learn More
MilliporeSigma™ Supelco™ Open-Top Screw Cap
Catalog No. 111011709 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Septum:
PTFE/Silicone
PTFE/Silicone/PTFE
No. per Pack:
100
1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum No. per Pack
11-101-1709 PTFE/Silicone 100
11-100-8889 PTFE/Silicone/PTFE 1000
11-100-9165 PTFE/Silicone 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-1709 Supplier MilliporeSigma™ Supelco™ Supplier No. 27326
Add to Cart
Add to Cart
Pre-assembled 9 mm polypropylene hole-cap, with septum.
  • Preassembled cap and septum.
  • Open-top polypropylene construction.
TRUSTED_SUSTAINABILITY
Specifications
Description Clear cap
Color Clear
Septum PTFE/Silicone
No. per Pack 100
Quantity 100 Ea.
Closure Size 9 mm