Learn More
MilliporeSigma™ Supelco™ Open-Top Screw Cap
Catalog No. 111011789 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Color:
Red, Tan
Catalog No. Color
11-101-1789 Red, Tan
1 options
Catalog No. 11-101-1789 Supplier MilliporeSigma™ Supelco™ Supplier No. 507415
Add to Cart
Add to Cart
Pre-assembled 9 mm polypropylene hole-cap, with septum.
TRUSTED_SUSTAINABILITY
Specifications
Color Red, Tan
Material Polypropylene
Septum PTFE/Silicone
No. per Pack 100
Closure Size 9 mm