Learn More
Bellco Glass, Inc. Open Top Screw Cap Only,15mm
SDP
Catalog No. 501945337
Encompass_Preferred
Dimensions: 0.7”OD x 0.38” thick, 0.343” hole. Size: 15mmMaterial: black phenolicThread finish: 15-425 . MADE IN THE USA
Catalog No. 50-194-5337 Supplier Bellco Glass, Inc. Supplier No. 204716000
Dimensions: 0.7”OD x 0.38” thick, 0.343” hole. Size: 15mmMaterial: black phenolicThread finish: 15-425 . MADE IN THE USA