Learn More
MilliporeSigma™ Supelco™ Open Top Screw Caps
Catalog No. 111011200 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Closure Size:
13-425
15-425
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
11-101-1200 15-425
11-101-1579 13-425
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-1200 Supplier MilliporeSigma™ Supelco™ Supplier No. Z106453200EA
Add to Cart
Add to Cart
Closure type screw cap with cut-out top to prevent contamination
  • Autoclavable
  • Easy sterilization
  • Used as a closure type for the containers used to contain substances in order to prevent contamination
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material Phenolic
Septum Without septum
No. per Pack 200
Closure Size 15-425