Learn More
MSC Value Collection Guidance Pulley, Malleable Iron, 1-3/8 in. Sheave, 5/16 in. Rope, 1 ea.
Catalog No. 5000512067
Encompass
This single open upright mount guidance pulley is designed for effective rope guidance in various applications. It features a robust construction suitable for use with fibrous rope, providing reliable performance in industrial or rigging setups.
  • Outside diameter of 1-3/8 in.
  • Compatible with fibrous rope up to 5/16 in. diameter
  • Constructed from durable malleable iron
  • Galvanized finish for enhanced corrosion resistance
  • Mounting hole diameter of 0.17 in. to 0.181 in.
Catalog No. 50-005-12067 Supplier MSC Supplier No. 67328401
This single open upright mount guidance pulley is designed for effective rope guidance in various applications. It features a robust construction suitable for use with fibrous rope, providing reliable performance in industrial or rigging setups.
  • Outside diameter of 1-3/8 in.
  • Compatible with fibrous rope up to 5/16 in. diameter
  • Constructed from durable malleable iron
  • Galvanized finish for enhanced corrosion resistance
  • Mounting hole diameter of 0.17 in. to 0.181 in.