Learn More
MSC Value Collection Single Open Upright Mount Guidance Pulley, Malleable Iron, 1-1/2 in. Sheave, 1 ea.
Catalog No. 5000510241
Encompass
The single open upright mount guidance pulley is designed for efficient rope management in various material handling applications. This pulley features a durable construction suitable for use with fibrous rope types, providing reliable support and guidance in overhead or mounting configurations.
  • Constructed from galvanized malleable iron for durability
  • Designed for use with 3/8 in. diameter fibrous rope
  • Includes a 1-1/2 in. outside diameter sheave
  • Features a mounting hole diameter of 0.209 in.
Catalog No. 50-005-10241 Supplier MSC Supplier No. 67328427
The single open upright mount guidance pulley is designed for efficient rope management in various material handling applications. This pulley features a durable construction suitable for use with fibrous rope types, providing reliable support and guidance in overhead or mounting configurations.
  • Constructed from galvanized malleable iron for durability
  • Designed for use with 3/8 in. diameter fibrous rope
  • Includes a 1-1/2 in. outside diameter sheave
  • Features a mounting hole diameter of 0.209 in.