Learn More
Opentrons Labworks Inc OPENTRONS 300UL TIP REFILLS
Catalog No. NC2071132
Encompass
  • Volume range: 5 µL to 300 µL
  • Tip material: Polypropylene
  • Tip color: Clear
  • Sterilization method: E-beam irradiation
  • Purity: DNase/RNase free, pyrogen free, protease free, human DNA free
  • Autoclavable: false
  • Compatible with: Opentrons OT-2 robotic system, Opentrons 8-Channel GEN2 P300 Electronic Pipettes, Opentrons Single-Channel GEN2 P300 Electronic Pipettes, Opentrons Single-Channel GEN1 P50 Electronic Pipettes, Opentrons Single-Channel GEN1 P300 Electronic Pipettes, Opentrons 8-Channel GEN1 P50 Electronic Pipettes, Opentrons 8-Channel GEN1 P300 Electronic Pipettes
  • Packaging quantity: 9,600 tips
  • Tip type: Non-filtered
Catalog No. NC2071132 Supplier Opentrons Labworks Inc Supplier No. 99900012
  • Volume range: 5 µL to 300 µL
  • Tip material: Polypropylene
  • Tip color: Clear
  • Sterilization method: E-beam irradiation
  • Purity: DNase/RNase free, pyrogen free, protease free, human DNA free
  • Autoclavable: false
  • Compatible with: Opentrons OT-2 robotic system, Opentrons 8-Channel GEN2 P300 Electronic Pipettes, Opentrons Single-Channel GEN2 P300 Electronic Pipettes, Opentrons Single-Channel GEN1 P50 Electronic Pipettes, Opentrons Single-Channel GEN1 P300 Electronic Pipettes, Opentrons 8-Channel GEN1 P50 Electronic Pipettes, Opentrons 8-Channel GEN1 P300 Electronic Pipettes
  • Packaging quantity: 9,600 tips
  • Tip type: Non-filtered