Learn More
Roboz Surgical Roboz Surgical Operating Scissors, Straight, Sharp-Sharp, 5.5 In, 1 Ea
SDP
Catalog No. NC0469795
Encompass
These 5.5-inch straight operating scissors are designed for precise tissue dissection. They feature one micro-serrated blade and one razor-honed blade, providing a smooth cutting action that requires minimal force while preventing tissue crushing or slippage.
  • Sharp-sharp tips for versatile use.
  • Micro-serrated edge prevents slippage on soft or tough tissue.
  • Bevel-honed razor edge minimizes tissue trauma.
  • Matte grey finger rings and black screw for identification.
  • Specifically engineered for cutting tissue only.
Catalog No. NC0469795 Supplier Roboz Surgical Supplier No. RS6814SC
These 5.5-inch straight operating scissors are designed for precise tissue dissection. They feature one micro-serrated blade and one razor-honed blade, providing a smooth cutting action that requires minimal force while preventing tissue crushing or slippage.
  • Sharp-sharp tips for versatile use.
  • Micro-serrated edge prevents slippage on soft or tough tissue.
  • Bevel-honed razor edge minimizes tissue trauma.
  • Matte grey finger rings and black screw for identification.
  • Specifically engineered for cutting tissue only.