Learn More
Med Vet International Operating Loupes, Premium, 3.5x Magnification, Short Working Distance, J1138PL, Each
Catalog No. 5000584265
Encompass_Preferred
Premium Loupes include case, cleaning cloth, flip paddle, lanyard and optical lens protector caps. Stylish sport frames with comfort gel nose bridge. One turn interpupillary adjustment. Premium Loupes, magnification 3.5x, Short 330mm working distance, depth of field 50mm, field of view 44mm, weight 2.9 oz, red frames.
Catalog No. 50-005-84265 Supplier Med Vet International Supplier No. JOR1138PL
Premium Loupes include case, cleaning cloth, flip paddle, lanyard and optical lens protector caps. Stylish sport frames with comfort gel nose bridge. One turn interpupillary adjustment. Premium Loupes, magnification 3.5x, Short 330mm working distance, depth of field 50mm, field of view 44mm, weight 2.9 oz, red frames.