Learn More
Hillrom Operating Otoscope
Catalog No. 22355142
Change view
Click to view available options
Voltage:
3.5 V
Catalog No. Voltage
22-355-142 3.5 V
1 options
This item is not returnable. View return policy
Catalog No. 22-355-142 Supplier Hillrom Supplier No. WEL 21700
Add to Cart
Add to Cart
This item is not returnable. View return policy
Operating Otoscope - Open system with rotatable lens and optional speculum for convenient procedures, plus bright light output.
  • Halogen HPX™ lamp for true tissue color and long-lasting performance
  • Open system makes use during procedures more convenient
  • Rotatable lens and speculum for ease of use
  • Fiber-optic illumination produces cool light
Specifications
Voltage 3.5 V
Disposable Reusable
Includes Reusable Ear Specula Set
Product Type Otoscope