Learn More
World Precision Instrument Operating Scissors, 14 cm, Straight, Blunt/Blunt, 12-pack
SDP
Catalog No. 502539450
Encompass
Ring-handled Operating Scissors are surgical scissors commonly used in general surgery for cutting and dissecting tissues. The 14 cm length is a versatile size that can be used for a variety of procedures, including biological and medical research. These operating scissors are available with straight or curved blades. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades. These are available in stainless steel. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. These economy scissors are sold in packages of 12, perfect for students or for disposable use. All sales are final on disposable instrument packs.
Catalog No. 50-253-9450 Supplier World Precision Instrument Supplier No. 50371712
Ring-handled Operating Scissors are surgical scissors commonly used in general surgery for cutting and dissecting tissues. The 14 cm length is a versatile size that can be used for a variety of procedures, including biological and medical research. These operating scissors are available with straight or curved blades. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades. These are available in stainless steel. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. These economy scissors are sold in packages of 12, perfect for students or for disposable use. All sales are final on disposable instrument packs.