Learn More
Sklar Instruments Operating Scissors, 5 1/2 in., Operating Scissors, Straight, Sharp/Blunt, EA.
SDP
Catalog No. 501109736 Shop All Sklar Instruments Products
Encompass_Preferred
Operating Scissors, 5 1/2 in., Operating Scissors, Straight, Sharp/Blunt, Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. This product is straight with smooth, sharp/blunt tips and a length of 5-1/2 inches.
Catalog No. 50-110-9736 Supplier Sklar Instruments Supplier No. 141055
Operating Scissors, 5 1/2 in., Operating Scissors, Straight, Sharp/Blunt, Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. This product is straight with smooth, sharp/blunt tips and a length of 5-1/2 inches.