Learn More
World Precision Instrument Operating Scissors, 16cm, Curved, Sharp/Blunt, 12-pack, Disposable
SDP
Catalog No. 502539449
Encompass
Ring-handled Operating Scissors are surgical scissors commonly used in general surgery for cutting and dissecting tissues. The 16 cm length is a versatile size that can be used for a variety of procedures, including biological and medical research. These operating scissors have curved blades, which are pointed on one blade and blunt on the other (sharp/blunt). Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These are available in stainless steel. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. These economy scissors are sold in packages of 12, perfect for students or for disposable use. All sales are final on disposable instrument packs.
Catalog No. 50-253-9449 Supplier World Precision Instrument Supplier No. 50371812
Ring-handled Operating Scissors are surgical scissors commonly used in general surgery for cutting and dissecting tissues. The 16 cm length is a versatile size that can be used for a variety of procedures, including biological and medical research. These operating scissors have curved blades, which are pointed on one blade and blunt on the other (sharp/blunt). Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These are available in stainless steel. Our Operating Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. These economy scissors are sold in packages of 12, perfect for students or for disposable use. All sales are final on disposable instrument packs.