Learn More
Sklar Instruments Operating Scissors, Sklar , 4-1/2 in., Standard, Straight, Sharp/Sharp, Smooth, Stainless Steel, Reusable. EA.
SDP
Catalog No. NC0393878 Shop All Sklar Instruments Products
Encompass_Preferred
Operating Scissors, Sklar, 4-1/2 in., Standard, Straight, Sharp/Sharp, Smooth, Stainless Steel, Reusable. Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. EA
Catalog No. NC0393878 Supplier Sklar Instruments Supplier No. 131045
Operating Scissors, Sklar, 4-1/2 in., Standard, Straight, Sharp/Sharp, Smooth, Stainless Steel, Reusable. Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. EA