Learn More
Sklar Instruments Econo Operating Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Straight, Smooth, Finger Rings, Stainless Steel. PK/12.
SDP
Catalog No. 501437575 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Operating Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Straight, Smooth, Finger Rings, Stainless Steel. Econo Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Econo instruments are, disposable, floor-grade instruments sold in bulk quantities. This product is straight with smooth, sharp/sharp tips and a length of 4-1/2 inches. They are sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7575 Supplier Sklar Instruments Supplier No. 21266
Econo Operating Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Straight, Smooth, Finger Rings, Stainless Steel. Econo Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Econo instruments are, disposable, floor-grade instruments sold in bulk quantities. This product is straight with smooth, sharp/sharp tips and a length of 4-1/2 inches. They are sold in boxes of 12.