Learn More
Sklar Instruments Operating Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel. Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument.
SDP
Catalog No. 501437357 Shop All Sklar Instruments Products
Encompass_Preferred
Operating Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel. Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp operating scissors cut tougher tissue and are frequently used for dissecting, whereas blunt operating scissors are more often used for cutting materials such as sutures, tubing, or gauze. This product is curved with smooth, sharp/sharp tips and a length of 4-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7357 Supplier Sklar Instruments Supplier No. 132045
Operating Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Curved, Smooth, Stainless Steel. Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp operating scissors cut tougher tissue and are frequently used for dissecting, whereas blunt operating scissors are more often used for cutting materials such as sutures, tubing, or gauze. This product is curved with smooth, sharp/sharp tips and a length of 4-1/2 inches.