Learn More
Sklar Instruments Sklarhone Operating Scissors, Sklar, 5-1/2 in., Sharp/Sharp, Curved, Razor/Smooth, Finger Rings / Dual Black, Stainless Steel. EA/1.
SDP
Catalog No. 501437377 Shop All Sklar Instruments Products
Encompass_Preferred
Sklarhone Operating Scissors, Sklar, 5-1/2 in., Sharp/Sharp, Curved, Razor/Smooth, Finger Rings / Dual Black, Stainless Steel. Supercut Sklarhone Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Supercut Sklarhone scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design.
This item is not returnable. View return policy
Catalog No. 50-143-7377 Supplier Sklar Instruments Supplier No. 153448
Sklarhone Operating Scissors, Sklar, 5-1/2 in., Sharp/Sharp, Curved, Razor/Smooth, Finger Rings / Dual Black, Stainless Steel. Supercut Sklarhone Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Supercut Sklarhone scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design.