Learn More
Sklar Instruments Surgi-OR Operating Scissors, Sklar 5-1/2 in., Sharp/Blunt, Straight, Smooth, Stainless Steel. EA.
SDP
Catalog No. 501184188 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR Operating Scissors, Sklar. 5-1/2 in., Sharp/Blunt, Straight, Smooth, Stainless Steel. Surgi-OR Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with smooth, sharp/blunt tips and a length of 5-1/2 inches.
Catalog No. 50-118-4188 Supplier Sklar Instruments Supplier No. 95272
Surgi-OR Operating Scissors, Sklar. 5-1/2 in., Sharp/Blunt, Straight, Smooth, Stainless Steel. Surgi-OR Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with smooth, sharp/blunt tips and a length of 5-1/2 inches.