Learn More
Roboz Surgical OPERATING SCISSORS CARBIDE
SDP
Catalog No. NC9029786
Encompass
These 5 1/2-inch straight operating scissors are designed for surgical and laboratory use, featuring durable tungsten carbide inserts for superior edge retention and long-term performance. The sharp-sharp tip configuration ensures precise tissue dissection and effective cutting capabilities.
  • Tungsten carbide blade inserts provide enhanced cutting performance and durability
  • Straight blade geometry allows for versatile surgical utility
  • Sharp-sharp tip design facilitates accurate tissue cutting and piercing
  • Balanced construction ensures stable handling during fine procedures
Catalog No. NC9029786 Supplier Roboz Surgical Supplier No. RS6834
These 5 1/2-inch straight operating scissors are designed for surgical and laboratory use, featuring durable tungsten carbide inserts for superior edge retention and long-term performance. The sharp-sharp tip configuration ensures precise tissue dissection and effective cutting capabilities.
  • Tungsten carbide blade inserts provide enhanced cutting performance and durability
  • Straight blade geometry allows for versatile surgical utility
  • Sharp-sharp tip design facilitates accurate tissue cutting and piercing
  • Balanced construction ensures stable handling during fine procedures